Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40551

Supelco

2-(2-Methoxyphenoxy)ethylamine hydrochloride monohydrate

analytical standard, pharmaceutical impurity standard

Synonym(s):

Carvedilol Related Compound E (USP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO2 · HCl · H2O
CAS Number:
Molecular Weight:
221.68
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0 wt. % water
≤5% related substances HPLC impurities
≤5.0% residual solvents

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C9H13NO2.ClH.H2O/c1-11-8-4-2-3-5-9(8)12-7-6-10;;/h2-5H,6-7,10H2,1H3;1H;1H2

InChI key

LDYFUCPSLVCTMR-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service