40519
4-Picoline
analytical standard
Synonym(s):
4-Methylpyridine
About This Item
grade
analytical standard
Quality Level
vapor density
3.2 (vs air)
vapor pressure
4 mmHg ( 20 °C)
assay
≥98.0% (GC)
autoignition temp.
1000 °F
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤0.5% water
refractive index
n20/D 1.504 (lit.)
n20/D 1.504-1.508
bp
145 °C (lit.)
mp
2.4 °C (lit.)
density
0.957 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
Cc1ccncc1
InChI
1S/C6H7N/c1-6-2-4-7-5-3-6/h2-5H,1H3
InChI key
FKNQCJSGGFJEIZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
102.2 °F - closed cup
flash_point_c
39 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service