Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39776

Supelco

Ibuprofen-(methyl-13C,d3)

analytical standard

Synonym(s):

α-Methyl-13C,d3-4-(isobutyl)phenylacetic acid, [13C,2H3]-Ibuprofen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC12D3H15O2
CAS Number:
Molecular Weight:
210.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

Storage temp.

2-8°C

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)/i3+1D3

Inchi Key

HEFNNWSXXWATRW-LBDFIVMYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magda Caban et al.
Analytical methods : advancing methods and applications, 12(30), 3854-3864 (2020-07-24)
The study aimed to show the limitations and advantages of the use of stable isotope labeled internal standards (SILISs) for the quantification of small polar compounds (ibuprofen, diclofenac, metoprolol, bisphenol A, 17β-estradiol) in water samples by GC/MS with selected ion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service