39278
Ethanol
puriss. p.a., ACS reagent, absolute alcohol, without additive, A15 o1, ≥99.8%
Synonym(s):
Ethyl alcohol
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
vapor density
1.59 (vs air)
vapor pressure
44.6 mmHg ( 20 °C)
assay
≥99.8%
form
liquid
autoignition temp.
683 °F
quality
absolute alcohol, without additive, A15 o1
expl. lim.
19 %, 60 °F
dilution
(for analytical testing)
impurities
KMnO4-reducing substances, in accordance
substances darkened by H2SO4, in accordance
≤0.0004% free alkali (as NH3)
≤0.001% aldehyde (as CH3CHO)
≤0.001% formaldehyde (HCHO)
≤0.001% ketone (as CH3COCH3)
≤0.002% free acid (as CH3COOH)
≤0.1% methanol (CH3OH)
≤0.2% water
evapn. residue
≤0.001%
refractive index
n20/D 1.3600 (lit.)
n20/D 1.362
bp
78 °C (lit.)
mp
−114 °C (lit.)
cation traces
Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.1 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg
SMILES string
CCO
format
neat
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Footnote
greener alternative product
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
55.4 °F - closed cup
flash_point_c
13 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service