Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39075

Supelco

Mercury ionophore I

Selectophore

Synonym(s):

7,16-Dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane, 1,10-Dibenzyl-1,10-diaza-18-crown-6, N,N′-Dibenzyl-4,13-diaza-18-crown-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H38N2O4
CAS Number:
Molecular Weight:
442.59
Beilstein/REAXYS Number:
1044804
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

description

for ion-selective electrodes

Quality Level

product line

Selectophore

form

crystals

mp

80-83 °C (lit.)

SMILES string

C1COCCN(CCOCCOCCN(CCO1)Cc2ccccc2)Cc3ccccc3

InChI

1S/C26H38N2O4/c1-3-7-25(8-4-1)23-27-11-15-29-19-21-31-17-13-28(24-26-9-5-2-6-10-26)14-18-32-22-20-30-16-12-27/h1-10H,11-24H2

InChI key

WAHZGOBRKWVALN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
It was used to coat two piezoelectric crystals required during the quantification of potassium.[1]

Other Notes

Highly selective ionophore for Hg2+ [2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The 1: 2 and 1: 1 molecular complexes of N, N'-dibenzyl-4, 13-diaza-18-crown-6 with 4-nitrobenzenesulfonamide and dithiooxamide.
Fonari, Marina S., et al.
Journal of Molecular Structure, 794, 110-114 (2006)
M Teresa et al.
The Analyst, 125(11), 1983-1986 (2001-02-24)
N,N'-dibenzyl-4,13-diaza-18-crown-6 (A) and bis[(benzo-15-crown-5)-4'-ylmethyl] pimelate (B) were tested as coatings for two piezoelectric crystals for potassium quantification. Both sensors showed stability, reversibility and sensitivity characteristics that allowed their use in quantitative analysis. However, compound A is much more sensitive to
V. K. Gupta et al.
Indian J. Chem. B, 42, 813 -813 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service