Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38835

Supelco

Bromide standard concentrate 10.00 gr Br-

analytical standard, 10.00 g/L, for 1 l standard solution

Synonym(s):

Potassium bromide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

contains

phenylmercury silver chloride as stabilizer

packaging

ampule of 14.893 g KBr

concentration

10.00 g/L

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Pan et al.
Organic letters, 13(18), 4974-4976 (2011-08-26)
A facile Pd-catalyzed conversion of aryl and vinyl triflates to aryl and vinyl halides (bromides and chlorides) is described. This method allows convenient access to a variety of aryl, heteroaryl, and vinyl halides in good to excellent yields and with
Kazue Kurihara
Advances in colloid and interface science, 158(1-2), 130-138 (2010-05-11)
Brush layers of polyelectrolytes, ionized chains of poly(glutamic acid) (PLGA) and poly(lysine) (PLL), prepared by the Langmuir-Blodgett method, were characterized using the surface forces measurements at various pHs, salt concentrations and chain densities. This paper reviews the major results: (1)
Keiichiro Hayashi et al.
Epilepsia, 52(5), 1010-1017 (2011-04-13)
Mutations in the SCN1A gene, which encodes the α1 subunit of voltage-gated sodium channels, cause generalized epilepsy with febrile seizures plus (GEFS+) and severe myoclonic epilepsy of infancy (SMEI). N1417H-Scn1a mutant rats are considered to be an animal model of
Pascal Miéville et al.
Magnetic resonance in chemistry : MRC, 49(11), 689-692 (2011-10-18)
As previously demonstrated by Thurber and Tycko, the peak position of (79)Br in potassium bromide (KBr) allows one to determine the temperature of a spinning sample. We propose to adapt the original design by using a compact KBr tablet placed
Thomas Trevethan et al.
Small (Weinheim an der Bergstrasse, Germany), 7(9), 1264-1270 (2011-04-13)
Modification and functionalization of the atomic-scale structure of insulating surfaces is fundamental to catalysis, self-assembly, and single-molecule technologies. Specially designed syn-5,10,15-tris(4-cyanophenylmethyl)truxene molecules can reshape features on an ionic KBr (001) surface. Atomic force microscopy images demonstrate that both KBr monolayer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service