Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
(CH3O)2C6H3CH2OH
CAS Number:
Molecular Weight:
168.19
EC Number:
202-212-0
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
639388
MDL number:
grade
purum
assay
≥95.0% (GC)
refractive index
n20/D 1.552 (lit.), n20/D 1.553
bp
296-297 °C/732 mmHg (lit.)
density
1.157 g/mL at 25 °C (lit.)
SMILES string
COc1ccc(CO)cc1OC
InChI
1S/C9H12O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-5,10H,6H2,1-2H3
InChI key
OEGPRYNGFWGMMV-UHFFFAOYSA-N
Other Notes
Protecting group for the guanosine residue during oligonucleotide synthesis
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
H. Takaku et al.
Chemical & Pharmaceutical Bulletin, 32, 2882-2882 (1984)
Liang Mao et al.
Environmental science & technology, 44(7), 2599-2604 (2010-03-17)
We have demonstrated in our earlier work that a few natural and synthetic estrogens can be effectively transformed through reactions mediated by lignin peroxidase (LiP). The behaviors of such reactions are variously influenced by the presence of natural organic matter
Takahisa Tsukihara et al.
Applied and environmental microbiology, 74(9), 2873-2881 (2008-03-11)
Unlike general peroxidases, Pleurotus ostreatus MnP2 was reported to have a unique property of direct oxidization of high-molecular-weight compounds, such as Poly R-478 and RNase A. To elucidate the mechanism for oxidation of polymeric substrates by MnP2, a series of