38229
3-Mercapto-3-methylbutyl formate
analytical standard
Synonym(s):
3-Mercapto-3-methyl-1-butyl-1-formate, Formic acid 3-mercapto-3-methylbutyl ester
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥97.5% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.460-1.462
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
storage temp.
2-8°C
SMILES string
CC(C)(S)CCOC=O
InChI
1S/C6H12O2S/c1-6(2,9)3-4-8-5-7/h5,9H,3-4H2,1-2H3
InChI key
VTAPYUYITKYXJB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Analysis Note
Recommended products
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
171.5 °F
flash_point_c
77.5 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service