Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

37885

Supelco

Fenthionsulfoxide

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15O4PS2
CAS Number:
Molecular Weight:
294.33
Beilstein/REAXYS Number:
2136026
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(c(C)c1)S(C)=O

InChI

1S/C10H15O4PS2/c1-8-7-9(5-6-10(8)17(4)11)14-15(16,12-2)13-3/h5-7H,1-4H3

InChI key

DLAPIMGBBDILHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonghwa Lee et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-26)
A simultaneous analytical method for the organophosphorus insecticide fenthion and its five metabolites (fenthion oxon, fenthion oxon sulfoxide, fenthion oxon sulfone, fenthion sulfoxide, and fenthion sulfone) was developed based on ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS). Five matrices (brown
Nasser Mokhtari et al.
Journal of the science of food and agriculture, 100(6), 2364-2371 (2019-12-20)
A new type of deep eutectic solvent based on three components using phosphate salts has been synthesized, characterized, and applied in the extraction of eight organothiophosphate pesticides from honey samples. In this study, the deep eutectic solvent was prepared from
Jishi Wang et al.
Journal of chromatography. A, 1521, 10-18 (2017-09-25)
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service