measuring range
0.1-10 mg/L
application(s)
environmental
general analytical
General description
Aquanal™ kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. AQUANAL™-professional tube tests were developed to provide a fast safe and clean method in order to perform water analysis in sewage treatment plants. The sample is pipetted into a tube that contains the test reagent. For some tests, an additional reagent is required that has to be added manually.
Packaging
24 reaction tubes, 1 blank tube
Analysis Note
Method: forming of a red-violet colored azo dye with N-(1-naphthyl)-ethylenediammonium dichloride
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service