application(s)
environmental
general analytical
General description
Aquanal™ kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. AQUANAL™-plus Thermoblock, provides reliable heating during sample preparation.
power unit; 8 boreholes 16 mm, 1 borehole 22 mm
Application
for heating of the AQUANAL™-plus tube tests
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)