usage
sufficient for 300 tests
measuring range
0.1-1.0 mg/L (photometric, Spectro 3)
application(s)
environmental
general analytical
General description
Aquanal™ kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations.
Analysis Note
Method: forming of a yellow-red colored fluorescent dye in a acidly medium
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 2
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Takeshi Tomita et al.
Journal of biochemistry, 164(6), 415-426 (2018-08-31)
A small nuclear protein, C1D, has roles in various cellular processes, transcription regulation, genome stability surveillance, DNA repair and RNA processing, all of which are required to maintain the host life cycles. In the previous report, C1D directly interacts with
Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service