Skip to Content
MilliporeSigma

37530

2,4-Dihydroxybenzoic acid

purum, ≥98.0% (T)

Synonym(s):

β-Resorcylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
EC Number:
201-946-9
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1946213
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥98.0% (T)

mp

208-211 °C (dec.) (lit.)

SMILES string

OC(=O)c1ccc(O)cc1O

InChI

1S/C7H6O4/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,8-9H,(H,10,11)

InChI key

UIAFKZKHHVMJGS-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 2,4-Dihydroxybenzoic acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Kamal Azrague et al.
Journal of hazardous materials, 237-238, 71-78 (2012-09-15)
2,4-Dihydroxybenzoic acid (2,4-DHBA) is found frequently as a pollutant in natural waters and represents a threat to water quality because it is a precursor to the formation of quinones which are highly toxic. The degradation of 2,4-DHBA using the vacuum
H A Schreuder et al.
Biochemistry, 33(33), 10161-10170 (1994-08-23)
The crystal structures of wild-type p-hydroxybenzoate hydroxylase from Pseudomonas fluorescens, complexed with the substrate analogues 4-aminobenzoate, 2,4-dihydroxybenzoate, and 2-hydroxy-4-aminobenzoate have been determined at 2.3-, 2.5-, and 2.8-A resolution, respectively. In addition, the crystal structure of a Tyr222Ala mutant, complexed with
A Stolz et al.
FEMS microbiology letters, 108(2), 219-224 (1993-04-01)
The aerobic degradation of 2,4-dihydroxybenzoate by Pseudomonas sp. BN9 was studied. Intact cells of Pseudomonas sp. BN9 grown with 2,4-dihydroxybenzoate oxidized 2,4-dihydroxybenzoate but not salicylate. Cell-free extracts of Pseudomonas sp. BN9 converted 2,4-dihydroxybenzoate after the addition of NAD(P)H. A partially



Global Trade Item Number

SKUGTIN
279390-1G04061837655876