application(s)
environmental
general analytical
Looking for similar products? Visit Product Comparison Guide
General description
Aquanal™ kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.
Application
AQUANAL™-plus nitrate (NO3) may be used as a calibrated standard during an experimental study conducted to study the mechanism of graphite oxide (GO) thermal reduction.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kinetic study of the graphite oxide reduction: combined structural and gravimetric experiments under isothermal and nonisothermal conditions.
Barroso B
The Journal of Physical Chemistry, 114 (49), 21645-21651 (2010)
Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service