37455
AQUANAL™-plus acid capacity 0.1 ml 1 mmol/L
alkalinity refill pack for 32017
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
41113319
application(s)
environmental
general analytical
Looking for similar products? Visit Product Comparison Guide
General description
Aquanal™ kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
signalword
Danger
hcodes
Hazard Classifications
Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2
Storage Class
3 - Flammable liquids
flash_point_f
57.2 °F
flash_point_c
14 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service