37434
AQUANAL™-plus peroxide (H2O2) 1-30 mg/L
test set with color comparator
About This Item
usage
sufficient for 180 tests
measuring range
1-30 mg/L (color chart)
1.0-100 mg/L (photometric, Spectro 3)
application(s)
environmental
general analytical
industrial hygiene
Looking for similar products? Visit Product Comparison Guide
General description
Analysis Note
Formation of a yellow color complex through oxidation to peroxide
Principle
DEV
Gradation (color chart)
0 - 1 - 2 - 3 - 5 - 7 - 10 - 15 - 20 - 30 mg/L (H2O2)
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class
8B - Non-combustible corrosive hazardous materials
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service