37425
AQUANAL™-plus aluminum (Al) 0.02-0.2 mg/L
test set with color comparator and check solution
About This Item
usage
sufficient for 200 tests
measuring range
0.01-0.5 mg/L (photometric, Spectro 3)
0.02-0.2 mg/L (color chart)
application(s)
environmental
general analytical
SMILES string
[Al]
InChI
1S/Al
InChI key
XAGFODPZIPBFFR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Analysis Note
Formation of a red-violet dye with chrom azurol S
Gradation (color chart)
0 - 0.02 - 0.035 - 0.05 - 0.07 - 0.1 - 0.13 - 0.16 - 0.2 mg/l
Legal Information
wgk_germany
nwg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service