Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37424

Supelco

AQUANAL-plus copper (Cu) 0.05-0.5 / 0.4-4.5 mg/L

test set with color comparator and check solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41113319

usage

sufficient for 75 tests

measuring range

0.005-5.0 mg/L (photometric, Spectro 3)
0.05-0.5 mg/dL (color chart, range 1)
0.4-4.5 mg/L (color chart, range 2)

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Analysis Note

Method
Creates a blue complex with cuprizon

Gradation (color charts)
0 - 0.05 - 0.08 - 0.12 - 0.16 - 0.20 - 0.25 - 0.30 - 0.40 - 0.50 mg/L (Cu2+, range 1)
0 - 0.4 - 0.7 - 1.0 - 1.4 - 1.8 - 2.2 - 2.8 - 3.5 - 4.5 mg/L (Cu2+, range 2)

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service