Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37416

Supelco

AQUANAL-plus total hardness 1 drop 1 German degree of hardness (0.178 mmol/L Ca)

titration test set

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41113319

usage

sufficient for ~500 tests

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Method: Complexometric titration
Principle: DEV

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service