Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37407

Supelco

AQUANAL-plus nickel (Ni) 0.02-0.5 mg/L

test set with color comparator and check solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41113319

usage

sufficient for 125 tests

measuring range

0.02-0.5 mg/L (color chart)
0.02-2.0 mg/L (photometric, Spectro 3)

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Method
Formation of a red-brown color complex with dimethylglyoxime

Principle
DEV

Gradation (color chart)
0 - 0.02 - 0.04 - 0.07 - 0.1 - 0.15 - 0.2 - 0.3 - 0.4 - 0.5 mg/L (Ni2+)

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1A Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT RE 2 Oral

target_organs

Lungs, Thyroid,Kidney,Liver,Blood

Storage Class

3 - Flammable liquids

flash_point_f

68.0 °F

flash_point_c

20 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service