Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37403

Supelco

AQUANAL-plus cyanide (CN) 0.03-0.7 mg/L

test set with color comparator

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41113319

usage

sufficient for 200 tests

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

measuring range

0.01-0.5 mg/L (photometric, Spectro 3)
0.03-0.7 mg/L (color chart)

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Method
Formation of a violet polymethine dye with pyridine and 1,3-dimethylbarbituric acid

Principle
DEV

Gradation (color chart)
0 - 0.03 - 0.06 - 0.10 - 0.15 - 0.20 - 0.30 - 0.45 - 0.70 mg/L (CN-)

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service