Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37213

Supelco

Sulfate test sticks Quantofix®

200-1600 mg/L (SO42-)

Synonym(s):

Quantofix® sulfate test sticks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

semi-quantitative

specific analyte(s)

sulfate

measuring range

200-1600 mg/L (SO42-)

application(s)

environmental
general analytical

detection method

colorimetric

General description

Contents:
Tube with 100 test sticks.

Storage:
Avoid exposing the strips to sunlight and moisture. Keep container cool and dry (store below +30 °C).

Gradation:
<200 · >400 · >800 · >1200 · >1600 mg/l SO42-

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service