Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37211

Supelco

Fluoride test kit

box of 30 discs (incl. reagent)

Synonym(s):

Fluoride test paper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

description

semi-quantitative

specific analyte(s)

fluoride

packaging

box of 30 discs (incl. reagent)

application(s)

environmental
food and beverages
general analytical

detection method

colorimetric

Application

The reaction is based on the bleaching action of fluoride ions on an aluminum color complex. The fluoride test is especially suited for rapid control of fluoride concentrations. Its purpose is to define concentration ranges and to detect values which fall short of or exceed given limits. Chlorates and bromates cause white discolorations of the test paper.

Analysis Note

Graduation
0 − 2 − 5 − 10 − 20 − 50 − 100 mg/l F

Color change
red to white

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service