Recommended Products
description
semi-quantitative
specific analyte(s)
chlorine
reaction suitability
reaction type: Redox Reactions
measuring range
1-100 mg/L (Cl2)
application(s)
environmental
food and beverages
general analytical
detection method
colorimetric
General description
Kit including reagents and test sticks for 100 tests.
Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.
Gradation:
0 - 1 - 3 - 10 - 30 - 100 mg/l Cl2
Legal Information
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
supp_hazards
Storage Class
3 - Flammable liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service