Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37202

Supelco

Potassium test kit Quantofix®

200-1500 mg/L (K+)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

semi-quantitative

specific analyte(s)

potassium

measuring range

200-1500 mg/L (K+)

application(s)

environmental
food and beverages
general analytical

detection method

colorimetric

General description

Contents:
Kit including reagents and test sticks for 100 tests.

Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.

Gradation:
0 - 200 - 400 - 700 - 1000 - 1500 mg/L K+

Application

It is used to measure extracellular potassium concentration using photometric method during potassium ion efflux assay.[1]

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trachyspermum ammi (L.) fruit essential oil influencing on membrane permeability and surface characteristics in inhibiting food-borne pathogens.
Paul, Souren, et al.
Food Control, 22, 725-731 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service