Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37144

Supelco

Litmus paper, neutral

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

feature

non-bleeding: no
refill: no

application(s)

agriculture
environmental
food and beverages
general analytical

detection method

colorimetric

Packaging

1 Package = 5 booklets with 100 strips each

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert L. Bonnet , Dan Keen and Frances W. Zweifel
Science Fair Projects: Chemistry, 12-12 (2000)
Chemical analyses of aqueous extract of Parkia biglobosa fruit husk collected from Northern Ghana.
Abagale, S. A., S. K. Twumasi, and J. A. M. Awudza.
Scientific Research and Essays, 8, 589-595 (2013)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 500-500 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service