Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37135

Supelco

Litmus paper blue

reag. Ph. Eur.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

agency

USP/NF
reag. Ph. Eur.

feature

non-bleeding: no
refill: no

application(s)

agriculture
environmental
food and beverages
general analytical

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

Packaging

1 Packing = 1 booklet with 100 strips each

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dr. K.R. Mahadik and Dr. S.H.Bhosale
Handbook of practical chemistry (Inorganic & organic), 164-164 (2008)
Iain E Yardley et al.
Clinical medicine (London, England), 10(3), 228-230 (2010-08-24)
Nasogastric tube insertion is a common clinical procedure carried out by doctors and nurses in NHS hospitals daily. For the last 30 years, there have been reports in the medical literature of deaths and other harm resulting from misplaced nasogastric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service