Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37111

Supelco

pH-Indicator strips

pH range 7.2-8.8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

feature

non-bleeding: no
refill: no

graduations and accuracy measuring range

accuracy: 0.2/0.3 pH unit

pH range

7.2-8.8

suitability

corresponds for shade of color acc. to scale

application(s)

general analytical

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

General description

Indicator papers are prepared by either direct dyes or reactive dyes. During reactive dyes procedure, pure linters are suspended in water and reactive dye is added. The mixture is made alkaline for the reaction to start. The fiber pulp is centrifuges, washed to be electrolyte-free and processed as pH paper. Paper produced in this way is bonded to plastic material in order to be used as stick or strip. Some strips are produced by impregnation which bleed, certain portion of the indicator dye dissolves in the test solution during immersion. pH strips with covalently bonded indicator dyes remain indefinitely in solution without migration or dissolution.[1]

Application

pH indicator strip may be used during the synthesis of modified nucleotides, for site-directed DNA photoaffinity labelling of RNA polymerase III transcription complexes.[2]

Features and Benefits

Each strip has an identification scale and printed pH scale. The strips can be utilized in colored solutions and suspensions with very high precision.

Packaging

Packages of 200 measuring strips.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Site-directed DNA photoaffinity labeling of RNA polymerase III transcription complexes.
J Persinger et al.
Methods in molecular biology (Clifton, N.J.), 148, 363-381 (2001-05-19)
Klaus Hunger
Industrial Dyes: Chemistry, Properties, Applications, 541-541 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service