Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37070

Supelco

Cyanide test kit Quantofix®

1-30 mg/L (CN-)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

description

semi-quantitative

specific analyte(s)

cyanide

measuring range

1-30 mg/L (CN-)

application(s)

environmental
food and beverages
forensics and toxicology
general analytical

detection method

colorimetric

General description

Contents:
Kit including reagents and test sticks for 100 tests.

Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.

Gradation:
0 − 1 − 3 − 10 − 30 mg/L CN-

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service