Recommended Products
description
semi-quantitative
specific analyte(s)
arsenic
measuring range
0.05-3.0 mg/L (As3+/5+)
application(s)
environmental
forensics and toxicology
general analytical
detection method
colorimetric
General description
Kit including reagents and test sticks for 100 tests.
Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.
Gradation:
0 − 0.05 − 0.1 − 0.5 − 1.0 − 1.7 − 3.0 mg/L As3+/5+
Legal Information
Disclaimer

signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service