37055
Special indicator paper
pH 0.5-5.5
Synonym(s):
Indicator paper, special
About This Item
Recommended Products
feature
non-bleeding: no
refill: no
pH
0.5-5.5
suitability
passes test for shade of color acc. to scale
application(s)
environmental
food and beverages
general analytical
industrial hygiene
detection method
colorimetric
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
roll with approx. 5 m
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service