Skip to Content
MilliporeSigma

37048

Sedaxane, mixture of isomers

PESTANAL®, analytical standard

Synonym(s):

Mixture of 2′-[(1RS,2RS)-1,1′-Bicycloprop-2-yl]-3-(difluoromethyl)-1-methylpyrazole-4-carboxanilide and 2′-[(1RS,2SR)-1,1′-bicycloprop-2-yl]-3-(difluoromethyl)-1-methylpyrazole-4-carboxanilide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H19F2N3O
CAS Number:
Molecular Weight:
331.36
UNSPSC Code:
41116107
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cn1cc(C(=O)Nc2ccccc2C3CC3C4CC4)c(n1)C(F)F

InChI

1S/C18H19F2N3O/c1-23-9-14(16(22-23)17(19)20)18(24)21-15-5-3-2-4-11(15)13-8-12(13)10-6-7-10/h2-5,9-10,12-13,17H,6-8H2,1H3,(H,21,24)

InChI key

XQJQCBDIXRIYRP-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
445401-5G04061832283821