Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37037

Supelco

Universal Indicator paper pH 1-11

triple color chart (refill pack with 3 rolls)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

Quality Level

feature

non-bleeding: no
refill: no

suitability

passes test for shade of color acc. to scale

application(s)

food and beverages
general analytical

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

Other Notes

this product will be discontinued, please find the alternative product here :
109527

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A precise method for the analysis of δ18O of dissolved inorganic phosphate in seawater.
McLaughlin, Karen, et al.
Limnology and Oceanography: Methods, 2, 202-212 (2004)
Denise Walker
Acids and Alkalis, 13-13 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service