37027
Special indicator paper
non-bleeding, pH 0.0-6.0
Synonym(s):
Indicator sticks, special
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41113319
description
coloring against scale complies
quality
non-bleeding
feature
non-bleeding: no
refill: no
pH
0.0-6.0
application(s)
environmental
food and beverages
general analytical
industrial hygiene
detection method
colorimetric
Looking for similar products? Visit Product Comparison Guide
General description
Special indicator papers cover range from 2 to 5 pH units in general. It has the capacity to change colour at particular pH value. Nonbleeding indicator papers are prepared by either direct dyes or reactive dyes. During reactive dyes procedure, pure linters are suspended in water and reactive dye is added. The mixture is made alkaline for the reaction to start. The fiber pulp is centrifuged, washed to be electrolyte-free and processed as pH paper. Paper produced in this way is bonded to plastic material in order to be used as a stick or strip.
Application
Special indicator paper may be used in volatile cyanide detection during Chitinase activity assay to understand the antifungal and phytostimulating characteristic of B. subtilis Ch-13 strain.
Packaging
box with 100 strips
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Antifungal and phytostimulating characteristics of Bacillus subtilis Ch-13 rhizospheric strain, producer of bioprepations.
Makarova, N. M., A. I. Shaposhnikov, and L. V. Kravchenko.
Applied Biochemistry and Microbiology, 45, 419-423 (2009)
Klaus Hunger
Industrial Dyes: Chemistry, Properties, Applications, 541-541 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service