Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37026

Supelco

pH-Universal-indicator sticks pH 0-14

non-bleeding

Synonym(s):

pH Indicator sticks, universal pH 0-14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

complies with coloring against scale

quality

non-bleeding

feature

non-bleeding: no
refill: no

application(s)

agriculture
environmental
food and beverages
general analytical
industrial hygiene

detection method

colorimetric

Packaging

box with 100 strips

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrically enhanced erosion of porous Si material in electrolyte by pH modulation and its application in chronotherapy.
Leong, W. Y., A. Loni, and L. T. Canham
Physica Status Solidi (A): Applied Research, 204, 1486-1490 (2007)
Denise Walker
Acids and Alkalis, 13-13 (2007)
Klaus Hunger
Industrial Dyes: Chemistry, Properties, Applications, 541-541 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service