Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37015

Supelco

Universal-Indicator paper (range pH 1.0-11.0; graduation 1.0 pH)

refill pack with 2×3 rolls

Synonym(s):

Universal indicator paper, pH 1,0-11,0

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

coloring against scale complies

feature

non-bleeding: no
refill: no

application(s)

food and beverages
general analytical

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A precise method for the analysis of δ18O of dissolved inorganic phosphate in seawater.
McLaughlin, Karen, et al.
Limnology and Oceanography: Methods, 2, 202-212 (2004)
Denise Walker
Acids and Alkalis, 13-13 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service