Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37014

Supelco

Universal-Indicator paper (range pH 1.0-11.0; graduation 1.0 pH)

Synonym(s):

Universal indicator paper, pH 1,0-11,0

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

description

complies with coloring against scale

feature

non-bleeding: no
refill: no

application(s)

food and beverages
general analytical

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

Packaging

6 boxes with 1 color chart
roll with approx. 5 m

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise Walker
Acids and Alkalis, 13-13 (2007)
A precise method for the analysis of δ18O of dissolved inorganic phosphate in seawater.
McLaughlin, Karen, et al.
Limnology and Oceanography: Methods, 2, 202-212 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service