Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36981

Supelco

Chlorothalonil solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8Cl4N2
CAS Number:
Molecular Weight:
265.91
Beilstein/REAXYS Number:
1978326
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1c(Cl)c(C#N)c(Cl)c(C#N)c1Cl

InChI

1S/C8Cl4N2/c9-5-3(1-13)6(10)8(12)7(11)4(5)2-14

InChI key

CRQQGFGUEAVUIL-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F

flash_point_c

2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin W King et al.
Environmental toxicology and chemistry, 32(3), 684-691 (2012-12-13)
Chlorothalonil concentrations exceeding acute toxicity levels for certain organisms have been measured in surface water discharge events from managed turf watersheds. The duration of exceedence and the timing of these events related to precipitation/runoff and time since application, however, have
Juan C Casas-Zapata et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(2), 122-132 (2013-01-12)
This study investigates the effects of chlorothalonil (CLT) on chemical oxygen demand (COD) and dissolved organic carbon (DOC) in pilot-scale horizontal subsurface flow constructed wetlands (HSSFCW) planted with Phragmites australis. Physicochemical parameters of influent and effluent water samples, microbial population
Christine Garron et al.
Pest management science, 68(1), 92-100 (2011-06-29)
Chlorothalonil is used extensively in Prince Edward Island for the control of blight on potatoes, and has been measured throughout summer months in ambient air monitoring studies. This study was designed to characterize near-field drift of chlorothalonil, and to evaluate
Md Musfiqur Rahman et al.
Biomedical chromatography : BMC, 27(4), 416-421 (2012-09-19)
A single extraction method was developed for chlorothalonil in shallot using gas chromatography with an electron capture detector (GC-μECD). Samples were extracted with single-step modified quick, easy, cheap, effective, rugged, and safe (QuEChERS) method using ethyl acetate as an extraction
Marie E DeLorenzo et al.
Marine pollution bulletin, 64(7), 1291-1299 (2012-06-12)
The precise application of risk assessment can lead to different conclusions about risk depending on how species are grouped in the assessment. We compared the use of different risk assessment methods for three different classes of pesticide, the herbicide diuron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service