Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36892

Supelco

cis-Permethrin solution

10 μg/mL in cyclohexane, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20Cl2O3
CAS Number:
Molecular Weight:
391.29
Beilstein/REAXYS Number:
9355109
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H](\C=C(\Cl)Cl)[C@H]1C(=O)OCc2cccc(Oc3ccccc3)c2

InChI

1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3/t17-,19-/m0/s1

InChI key

RLLPVAHGXHCWKJ-HKUYNNGSSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-0.4 °F

flash_point_c

-18 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temitope O Alimi et al.
Journal of the American Mosquito Control Association, 29(1), 49-53 (2013-05-22)
The efficacy of a new water-based formulation containing 30% permethrin and 30% piperonyl butoxide against laboratory and field populations of Aedes albopictus was evaluated in the laboratory, in semifield experiments, and in residential communities in St. Augustine, FL. In laboratory
Raweewan Srisawat et al.
The Southeast Asian journal of tropical medicine and public health, 43(6), 1380-1386 (2013-02-19)
One of the mechanisms responsible for pyrethroid resistance in mosquitoes is mutations in domain IIS6 of voltage-gated sodium channel gene (kdr). Aedes aegypti larvae were collected from the central provinces of Thailand (Bangkok, Prachin Buri and Ratchaburi) and colonized until
Juan Du et al.
Environmental toxicology and chemistry, 32(6), 1403-1411 (2013-02-21)
Compared with acute toxicity, chronic exposures to low levels of contaminants are more environmentally relevant, but fewer data are available. In the present study, sediment toxicity of the pyrethoid permethrin to Chironomus dilutus was determined. The whole-life-cycle toxicity testing was
Ian F Burgess et al.
BMC dermatology, 13, 5-5 (2013-04-04)
A previous study indicated that a single application of 4% dimeticone liquid gel was effective in treating head louse infestation. This study was designed to confirm this in comparison with two applications of 1% permethrin. We have performed a single
Michael Leschnik et al.
Parasites & vectors, 6, 62-62 (2013-03-19)
Tick-borne infections resulting from regular tick infestation in dogs are a common veterinary health problem all over the world. The application of repellent and acaricidal agents to prevent transmission of pathogens is a major protection strategy and has been proven

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service