Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

36734

Supelco

Phthalimide

PESTANAL®, analytical standard

Synonym(s):

1,3-Dihydro-1,3-dioxoisoindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO2
CAS Number:
Molecular Weight:
147.13
Beilstein/REAXYS Number:
118522
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

232-235 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

O=C1NC(=O)c2ccccc12

InChI

1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-4H,(H,9,10,11)

InChI key

XKJCHHZQLQNZHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Ion Mobility: TWCCSN2 value of 124.9 Å2 [M+H]+ and 124.0 Å2 [M-H]-

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 36734 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Abhishek A Kantak et al.
Journal of the American Chemical Society, 133(49), 19960-19965 (2011-10-21)
The development of a novel intermolecular oxidative amination reaction, a synthetic transformation that involves the simultaneous functionalization of both a N-H and C-H bond, is described. The process, which is mediated by an I(III) oxidant and contains no metal catalysts
Supramolecular chemistry: Reading polymer codes.
Ken D Shimizu
Nature chemistry, 2(8), 612-613 (2010-07-24)
Hülya Akgün et al.
Bioorganic & medicinal chemistry, 20(13), 4149-4154 (2012-05-29)
Structurally modified phthalimide derivatives were prepared through condensation of phthalic and tetrafluorophthalic anhydride with selected sulfonamides with variable yields. All compounds were screened for their antimycobacterium activity against Mycobacterium tuberculosis H37Ra (ATCC 25177) using a micro broth dilution technique. The
Baocun Zhu et al.
Organic & biomolecular chemistry, 8(7), 1650-1654 (2010-03-20)
A highly selective ratiometric fluorescent probe, which contains an aminonaphthalimide fluorophore and a self-immolative spacer for 1,4-dithiothreitol (DTT) detection was designed and synthesized. The probe displays a 66 nm red-shift of fluorescence emission and the color changes from colorless to
Zhengfang Wang et al.
Journal of the American Society for Mass Spectrometry, 23(3), 520-529 (2012-01-10)
We previously reported that selenamide reagents such as ebselen and N-(phenylseleno)phthalimide (NPSP) can be used to selectively derivatize thiols for mass spectrometric analysis, and the introduced selenium tags are useful as they could survive or removed with collision-induced dissociation (CID).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service