Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36693

Supelco

2-Chloroethanol

PESTANAL®, analytical standard

Synonym(s):

Ethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2OH
CAS Number:
Molecular Weight:
80.51
Beilstein/REAXYS Number:
878139
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor density

2.78 (vs air)

vapor pressure

5 mmHg ( 20 °C)

product line

PESTANAL®

autoignition temp.

797 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

16 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.441 (lit.)

bp

129 °C (lit.)

mp

−89 °C (lit.)

solubility

H2O: miscible (completely)

density

1.201 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OCCCl

InChI

1S/C2H5ClO/c3-1-2-4/h4H,1-2H2

InChI key

SZIFAVKTNFCBPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

131.0 °F - closed cup

flash_point_c

55 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Bednar et al.
Journal of hazardous materials, 168(2-3), 1041-1046 (2009-04-07)
Active treatment of BCEE (bis-2-chloroethyl ether) is being currently performed in the on-site Cohansey Aquifer at the Lipari Superfund Site. Remediation of BCEE in the underlying Kirkwood aquifer is being considered, necessitating investigations of BCEE geochemistry in aquifer material from
John W Barton et al.
Chemosphere, 73(11), 1737-1740 (2008-10-22)
We report measurements of solubility limits for benzene, toluene, and TCE in systems that contain varying levels of biomass up to 0.13 g mL(-1) for TCE and 0.25 g mL(-1) for benzene and toluene. The solubility limit increased from 21
Ernest Marco-Urrea et al.
Chemosphere, 70(3), 404-410 (2007-08-19)
The white-rot fungus Trametes versicolor degraded trichloroethylene (TCE), a highly oxidized chloroethene, and produced 2,2,2-trichloroethanol and carbon dioxide as the main products of degradation, based on the results obtained using [13C]-TCE as the substrate. For a range of concentrations of
Ronan Garlantézec et al.
Occupational and environmental medicine, 69(1), 62-70 (2011-06-28)
To describe urine levels of metabolites of glycol ethers and chlorinated solvents in a sample of pregnant women from the general population, to study their occupational and non-occupational determinants and to compare them with the results of indirect assessment methods
Mariella Carrieri et al.
Archives of toxicology, 81(7), 529-532 (2007-02-08)
Nonfatal acute inhalation of trichloroethylene (TRI) at work was described. The subject, male, 54 years old, was drawn unconscious by a metal-degreasing machine and immediately sheltered in intensive care unit. Other than basic life support and common laboratory indices, blood

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service