Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36673

Supelco

Endosulfan ether

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6Cl6O
CAS Number:
Molecular Weight:
342.86
Beilstein/REAXYS Number:
22311
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[H][C@@]12COC[C@]1([H])C3(Cl)C(Cl)=C(Cl)C2(Cl)C3(Cl)Cl

InChI

1S/C9H6Cl6O/c10-5-6(11)8(13)4-2-16-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3-,4+,7?,8?

InChI key

CKPWHXBGVRURFU-NYQGKTAPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingfen Kong et al.
Ecotoxicology and environmental safety, 160, 75-83 (2018-05-25)
The recently discovered endosulfan-degrading bacterial strains Pusillimonas sp. JW2 and Bordetella petrii NS were isolated from endosulfan-polluted water and soil environments. The optimal conditions for the growth and biodegradation activity of the strains JW2 and NS were studied in detail.
Murat Ozdal et al.
3 Biotech, 7(2), 113-113 (2017-06-02)
Stenotrophomonas maltophilia OG2 was isolated from the intestine of cockroaches that was collected from a cow barn contaminated some pesticides belong to pyrethroid and organochlorine groups. OG2 was able to degrade α-endosulfan in non sulfur medium (NSM) as a sole

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service