Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36605

Supelco

2,6-Dichlorobenzamide

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CONH2
CAS Number:
Molecular Weight:
190.03
Beilstein/REAXYS Number:
1869103
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

196-199 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

NC(=O)c1c(Cl)cccc1Cl

InChI

1S/C7H5Cl2NO/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)

InChI key

JHSPCUHPSIUQRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dichlorobenzamide is found to be a degradation product and metabolite[1] of the pesticide dichlobenil.[2][3][4]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian R Sørensen et al.
Applied and environmental microbiology, 73(2), 399-406 (2006-11-23)
2,6-Dichlorobenzamide (BAM), a persistent metabolite from the herbicide 2,6-dichlorobenzonitrile (dichlobenil), is the pesticide residue most frequently detected in Danish groundwater. A BAM-mineralizing bacterial community was enriched from dichlobenil-treated soil sampled from the courtyard of a former plant nursery. A BAM-mineralizing
Erik Belleville et al.
Journal of immunological methods, 286(1-2), 219-229 (2004-04-17)
To replace a pesticide immunoassay based on microtiter plates, we have developed a quantitative, competitive microarray immunoassay, which permits rapid and highly sensitive quantification of the dichlobenil degradation product 2,6-dichlorobenzamide (BAM), and the prominently used herbicide atrazine. The pesticide analysis
Filippo G Bosco et al.
Lab on a chip, 11(14), 2411-2416 (2011-05-31)
Sensors are crucial in many daily operations including security, environmental control, human diagnostics and patient monitoring. Screening and online monitoring require reliable and high-throughput sensing. We report on the demonstration of a high-throughput label-free sensor platform utilizing cantilever based sensors.
Degradation and mineralization of nanomolar concentrations of the herbicide dichlobenil and its persistent metabolite 2, 6-dichlorobenzamide by Aminobacter spp. isolated from dichlobenil-treated soils
S?rensen.RS, et al.
Applied and Environmental Microbiology, 73, 399-406 (2007)
Genetic (In) stability of 2, 6-Dichlorobenzamide Catabolism in Aminobacter sp. Strain MSH1 Biofilms under Carbon Starvation Conditions
Horemans B, et al.
Applied and Environmental Microbiology, 83 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service