Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36569

Supelco

Plifenat

PESTANAL®, analytical standard

Synonym(s):

2,2,2-Trichloro-1-(3,4-dichlorophenyl)ethyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Cl5O2
CAS Number:
Molecular Weight:
336.43
Beilstein/REAXYS Number:
6294075
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(=O)OC(c1ccc(Cl)c(Cl)c1)C(Cl)(Cl)Cl

InChI

1S/C10H7Cl5O2/c1-5(16)17-9(10(13,14)15)6-2-3-7(11)8(12)4-6/h2-4,9H,1H3

InChI key

FSGNOVKGEXRRHD-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Mäntylä et al.
Toxicology, 27(3-4), 327-336 (1983-07-01)
The effects of 2,2,2-trichloro-1-(3,4-dichlorophenyl)ethyl acetate (Penfenate) on hepatic, renal and small intestinal drug-metabolizing enzyme activities, hepatic reduced glutathione content and urinary excretion of thioethers were studied in the rat. A single i.p. dose of Penfenate (500 mg/kg) decreased the body
Miscellaneous notes on the toxicity of some insecticides for the head louse, Pediculus capitis De Geer (Anoplura).
L Blommers et al.
Acta Leidensia, 46, 9-15 (1978-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service