Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36548

Supelco

Methyl trichloroacetate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3CO2CH3
CAS Number:
Molecular Weight:
177.41
Beilstein/REAXYS Number:
1756075
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.455 (lit.)

bp

152-153 °C (lit.)

density

1.488 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)C(Cl)(Cl)Cl

InChI

1S/C3H3Cl3O2/c1-8-2(7)3(4,5)6/h1H3

InChI key

VHFUHRXYRYWELT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Candiano et al.
Analytical biochemistry, 243(2), 245-248 (1996-12-15)
This paper describes a new, sensitive (in the nanogram range), and rapid (two-step) technique for the negative staining of proteins in polyacrylamide gels in the presence or absence of sodium dodecyl sulfate. After separation, gels are incubated with 8% methyl
R Bansal et al.
Arzneimittel-Forschung, 62(9), 420-424 (2012-07-14)
In this study, O-alkylated derivatives of nafimidone oxime chemically, 1-(2-naphthyl)-2-(imidazol-1-yl)ethanone oxime have been synthesized as potential anticonvulsant compounds. O-alkylation of the oxime using hydrochlorides of various dialkylaminoethyl chlorides, methyl chloroacetate and alkyl dihalides gave the O-alkylated derivatives. Anticonvulsant activity of
Tianlin Wang et al.
Journal of chromatography. A, 1147(1), 105-110 (2007-03-03)
This paper reports the use of methyl chloroacetate (MCA) as an extraction solvent for coupling liquid-liquid semimicroextraction (LLsME) with micellar electrokinetic chromatography (MEKC) through on-capillary decomposition for the separation of neutral compounds with concentration enhancement. Alkylphenones of C(8), C(10) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service