Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36547

Supelco

Methyl dichloroacetate

PESTANAL®, analytical standard

Synonym(s):

Dichloroacetic acid methyl ester, NSC 2882

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHCOOCH3
CAS Number:
Molecular Weight:
142.97
Beilstein/REAXYS Number:
1747542
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.442 (lit.)

bp

143 °C (lit.)

mp

−52 °C (lit.)

density

1.381 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)C(Cl)Cl

InChI

1S/C3H4Cl2O2/c1-7-3(6)2(4)5/h2H,1H3

InChI key

HKMLRUAPIDAGIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes
  • Azide-alkyl halide cycloaddition and atom transfer radical addition
  • Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines
  • Three component reaction of arynes with cyclic ethers and active methines
  • Catalytic radical haloalkylation for synthesis of neodysidenin
  • Addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

147.2 °F - closed cup

flash_point_c

64 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service