Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36511

Supelco

Sebuthylazin-desethyl

PESTANAL®, analytical standard

Synonym(s):

Desethylsebutylazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12ClN5
CAS Number:
Molecular Weight:
201.66
Beilstein/REAXYS Number:
783753
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCC(C)Nc1nc(N)nc(Cl)n1

InChI

1S/C7H12ClN5/c1-3-4(2)10-7-12-5(8)11-6(9)13-7/h4H,3H2,1-2H3,(H3,9,10,11,12,13)

InChI key

VMYDDULRGBGGBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sebuthylazin-desethyl is commonly used as an analytical reference standard for environmental and food residue analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service