Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36394

Supelco

Maize GMO Standard MON 863

Set for 0%, 0.1%, 1% and 9,9% MON 863

Synonym(s):

GMO Maize Powder Standard MON 863

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.

product line

BioChemika

application(s)

cleaning products
cosmetics
food and beverages
microbiology
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service