Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36327

Supelco

(−)-β-Pinene

reference material

Synonym(s):

(1S,5S)-2(10)-Pinene, (1S,5S)-6,6-Dimethyl-2-methylenebicyclo[3.1.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein/REAXYS Number:
2038282
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

assay

≥98.5% (GC)

optical activity

[α]/D -22.0±1.0°, neat

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.477-1.479

bp

164-165 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC1(C)[C@H]2CCC(=C)[C@@H]1C2

InChI

1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9-/m0/s1

InChI key

WTARULDDTDQWMU-IUCAKERBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

102.2 °F - closed cup

flash_point_c

39 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service