Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36271

Supelco

4,5-Dimethoxy-1,2-phenylenediamine hydrochloride

LiChropur, ≥98.0% (GC)

Synonym(s):

1,2-Diamino-4,5-dimethoxybenzene hydrochloride, 4,5-Dimethoxybenzene-1,2-diamine hydrochloride, DDB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O2 · 2HCl
CAS Number:
Molecular Weight:
241.11
Beilstein/REAXYS Number:
14573971
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((HPLC))

Quality Level

assay

≥98.0% (GC)

quality

LiChropur

technique(s)

HPLC: suitable

storage temp.

−20°C

InChI

1S/C8H12N2O2/c1-11-7-3-5(9)6(10)4-8(7)12-2/h3-4H,9-10H2,1-2H3

InChI key

SKIUVOVOIJBJPN-UHFFFAOYSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorimetric determination of aromatic aldehydes with 4,5-dimethoxy-1,2-diaminobenzene.
Nakamura, M., et al.
Analytica Chimica Acta, 134, 39-45 (1982)
Fluorescent product in the determination of aromatic aldehydes with 4,5-dimethoxy-1,2-diaminobenzene.
Nakamura, M., et al.
Chemical & Pharmaceutical Bulletin, 31, 2910-2912 (1983)
S Hara et al.
Journal of chromatography, 377, 111-119 (1986-04-25)
A simple, rapid and highly sensitive high-performance liquid chromatographic method for the determination of N-acetyl- and N-glycolylneuraminic acids in serum and urine is described. The neuraminic acids, released by hydrolysis of serum and urine, are converted in dilute sulphuric acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service